Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125394.1 | 3 - 4 | 722 | 72 | ||
| GenBank mRNA | AK075529.1 | - | 3 - 4 | 625 | 72 | |
| BM549126.1 | - | 3 - 4 | 559 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 34C | ANKRD34C | -16.3kb | Sense | Chr. 15 - 79282804 - 79298239 on NCBI Build GRCh38 | |
| Downstream | KIAA1024 | KIAA1024 | 90.6kb | Sense | Chr. 15 - 79411557 - 79472304 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000561560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157558.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157558.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157558.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157558.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157558.hg.1 | TC1500008003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157560.hg.1 | TC1500008003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157562.hg.1 | TC1500008003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000561560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |

TMED3
Genomic Map