Assay Details
| Gene Symbol: LOC101927915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125428.1 | 2 - 3 | 376 | 73 | ||
| GenBank mRNA | EF565097.1 | - | 2 - 3 | 377 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
KH RNA binding domain containing, signal transduction associated 3 |
KHDRBS3 |
Etle, SALP, SLM-2, SLM2, T-STAR, TSTAR, etoile |
-1.7mb | Anti-sense | Chr. 8 - 135457461 - 135656516 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC401478 | LOC401478 | 405.7kb | Sense | Chr. 8 - 137812895 - 138083545 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927915
Genomic Map