Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
ENST00000431686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
ENST00000431686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150210.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150210.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150210.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150210.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150210.hg.1 |
TC1300007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300150215.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150215.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150215.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150215.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150215.hg.1 |
TC1300007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
ENST00000431686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
ENST00000431686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
ENST00000433664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007439.hg.1 |
NR_125752.1 |
Refseq |