Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009546.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165353.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165353.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165353.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165353.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165353.hg.1 |
TC0900009546.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165354.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165354.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165354.hg.1 |
TC0900009546.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165352.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165352.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165352.hg.1 |
ENST00000417638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165352.hg.1 |
NR_125775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165352.hg.1 |
TC0900009546.hg.1 |
Clariom D TC |