Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180969.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180969.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180969.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180969.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180969.hg.1 |
TC0100011035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100180970.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180970.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180970.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180970.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180970.hg.1 |
TC0100011035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011035.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180971.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180971.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180971.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180971.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180971.hg.1 |
TC0100011035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100180972.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180972.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180972.hg.1 |
ENST00000420807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180972.hg.1 |
NR_125789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180972.hg.1 |
TC0100011035.hg.1 |
Clariom D TC |