Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222433.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222433.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222433.hg.1 |
TC0200016478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016478.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
ENST00000377469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
ENST00000418204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
ENST00000435984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
NR_125792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222438.hg.1 |
TC0200016478.hg.1 |
Clariom D TC |