Assay Details
Gene Symbol: ZFPM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125796.1 | 4 - 5 | 442 | 71 | ||
GenBank mRNA | BF977562.1 | - | 329 | 71 | ||
BU959299.1 | - | 417 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LDL receptor related protein 12 | LRP12 | ST7 | -1.2mb | Sense | Chr. 8 - 104489231 - 104589024 on NCBI Build GRCh38 |
Downstream | oxidation resistance 1 | OXR1 |
Nbla00307, TLDC3 |
482.6kb | Anti-sense | Chr. 8 - 106270178 - 106752694 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177764.hg.1 | TC0800011417.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000518932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000520594.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | ENST00000524045.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | NR_125796.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177765.hg.1 | TC0800011417.hg.1 | Clariom D TC |
