Assay Details
Gene Symbol: LOC101929294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125808.1 | 1 - 2 | 75 | 91 | ||
GenBank mRNA | DA221074.1 | - | 1 - 2 | 75 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929315 | LOC101929315 | Sense | Chr. 8 - 24490312 - 24514868 on NCBI Build GRCh38 | ||
Downstream |
neurofilament, medium polypeptide |
NEFM |
NEF3, NF-M, NFM |
365.1kb | Anti-sense | Chr. 8 - 24913761 - 24919093 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800167232.hg.1 | TC0800009897.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000518988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000519689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | ENST00000523700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009897.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | ENST00000523578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | NR_125808.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800167233.hg.1 | TC0800009897.hg.1 | Clariom D TC |
