Assay Details
| Gene Symbol: LOC101929897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125823.1 | 2 - 3 | 455 | 64 | ||
| GenBank mRNA | AI025346.1 | - | 1 - 2 | 64 | ||
| BX094193.1 | - | 2 - 3 | 227 | 64 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  3' Most  Recommended Assay  Homo sapiens  Probe spans exons | 
MicroArray Probes:
					
						
							Affymetrix
					
					
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521470.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521470.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518213.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000518994.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000520890.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521470.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521470.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000521802.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | ENST00000523459.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0800010278.hg.1 | NR_125823.1 | Refseq | 
 Back To Top
 Back To Top
 LOC101929897
LOC101929897 Genomic Map
Genomic Map Back To Top
 Back To Top