Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175576.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175576.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175576.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175576.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175576.hg.1 |
TC0800011102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
ENST00000523284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011102.hg.1 |
NR_125827.1 |
Refseq |