Assay Details
Gene Symbol: LOC101928231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125852.1 | 1 - 2 | 689 | 114 | ||
GenBank mRNA | BC040308.1 | - | 1 - 2 | 691 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1010 | LINC01010 | Anti-sense | Chr. 6 - 134437716 - 134504020 on NCBI Build GRCh38 | ||
Downstream | uncharacterized LOC101928304 | LOC101928304 | 46.4kb | Sense | Chr. 6 - 134525318 - 134540005 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
