Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202613.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202613.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202613.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202613.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202613.hg.1 |
TC0600014340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
ENST00000422736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014340.hg.1 |
NR_125852.1 |
Refseq |