Assay Details
| Gene Symbol: LOC102724053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125873.1 | 2 - 3 | 389 | 119 | ||
| GenBank mRNA | AA437152.1 | - | 2 - 3 | 388 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibronectin type III domain containing 1 | FNDC1 |
AGS8, FNDC2, MEL4B3, bA243O10.1, dJ322A24.1 |
-121.1kb | Sense | Chr. 6 - 159169397 - 159272108 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC401281 | LOC401281 | 192.0kb | Anti-sense | Chr. 6 - 159588439 - 159589980 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102724053
Genomic Map