Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000511346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000511346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191452.hg.1 |
TC0400012995.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000511346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000511346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012995.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
ENST00000503140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
ENST00000515444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
NR_125896.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191443.hg.1 |
TC0400012995.hg.1 |
Clariom D TC |