Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169238.hg.1 |
TC0400010103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010103.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
ENST00000513384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169239.hg.1 |
TC0400010103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169242.hg.1 |
TC0400010103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
ENST00000502344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
NR_125911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169243.hg.1 |
TC0400010103.hg.1 |
Clariom D TC |