Assay Details
| Gene Symbol: BHLHE40-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125915.1 | 1 - 2 | 166 | 103 | ||
| GenBank mRNA | AK056892.1 | - | 1 - 2 | 166 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | basic helix-loop-helix family member e40 | BHLHE40 |
BHLHB2, DEC1, HLHB2, SHARP-2, STRA13, Stra14 |
Anti-sense | Chr. 3 - 4979412 - 4985181 on NCBI Build GRCh38 | |
| Downstream | ADP ribosylation factor like GTPase 8B | ARL8B |
ARL10C, Gie1 |
142.3kb | Anti-sense | Chr. 3 - 5122245 - 5180916 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173900.hg.1 | TC0300010139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173901.hg.1 | TC0300010139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000420832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000434530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000438479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000441386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000615178.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | ENST00000620618.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_037903.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010139.hg.1 | NR_125916.1 | Refseq |

BHLHE40-AS1
Genomic Map