Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169609.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169609.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169609.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169609.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169609.hg.1 |
TC0400010156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010156.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169611.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169611.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169611.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169611.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169611.hg.1 |
TC0400010156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400169612.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169612.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169612.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169612.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169612.hg.1 |
TC0400010156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400169610.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169610.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169610.hg.1 |
ENST00000509964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400169610.hg.1 |
NR_125919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400169610.hg.1 |
TC0400010156.hg.1 |
Clariom D TC |