Assay Details
Gene Symbol: LOC256880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125923.1 | 1 - 2 | 185 | 77 | ||
GenBank mRNA | DB462285.1 | - | 1 - 2 | 156 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | H2A histone family member Z | H2AFZ |
H2A.Z-1, H2A.z, H2A/z, H2AZ |
-139.0 | Anti-sense | Chr. 4 - 99948087 - 99950355 on NCBI Build GRCh38 |
Downstream | DNA damage inducible transcript 4 like | DDIT4L |
REDD2, Rtp801L |
159.8kb | Anti-sense | Chr. 4 - 100185870 - 100190498 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156766.hg.1 | TC0400008244.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156760.hg.1 | TC0400008244.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156759.hg.1 | TC0400008244.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000501976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000507494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | ENST00000515026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | NR_125923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008244.hg.1 | NR_125923.1 | Refseq |
