Assay Details
| Gene Symbol: MIR558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030285.1 | 1 | 48 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Yip1 domain family member 4 | YIPF4 |
FinGER4, Nbla11189 |
-224.8kb | Sense | Chr. 2 - 32277889 - 32307929 on NCBI Build GRCh38 |
| Downstream | BIRC6 antisense RNA 2 | BIRC6-AS2 |
linc-birc6, megamind |
25.3kb | Anti-sense | Chr. 2 - 32557520 - 32574818 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200007191.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000214.hg.1
Set Membership: |
|

MIR558
Genomic Map