Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007913.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155685.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155685.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155685.hg.1 |
ENST00000415255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155685.hg.1 |
NR_125979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155685.hg.1 |
TC0100007913.hg.1 |
Clariom D TC |