Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017237.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000412221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000431307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
ENST00000443448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
NR_125983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
NR_125984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230526.hg.1 |
TC0100017237.hg.1 |
Clariom D TC |