Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018541.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018541.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018541.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018541.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018541.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244263.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244263.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244263.hg.1 |
TC0100018541.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018541.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018541.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018541.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018541.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018541.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244265.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244265.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244265.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244265.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244265.hg.1 |
TC0100018541.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100244271.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244271.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244271.hg.1 |
ENST00000451690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244271.hg.1 |
NR_126003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244271.hg.1 |
TC0100018541.hg.1 |
Clariom D TC |