Assay Details
Gene Symbol: LOC101927973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126012.1 | 3 - 4 | 524 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC284825 | LOC284825 | -32.5kb | Anti-sense | Chr. 21 - 28048414 - 28137609 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 161 | LINC00161 |
C21orf100, NCRNA00161 |
327.0kb | Anti-sense | Chr. 21 - 28539318 - 28540355 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152835.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152837.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152817.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152821.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152824.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152831.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152834.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152819.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | TC2100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152826.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000519703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000520192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | ENST00000628324.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011889.hg.1 | NR_125774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
