Assay Details
Gene Symbol: SAP30BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126037.1 | 6 - 7 | 747 | 112 | ||
GenBank mRNA | AK298060.1 | - | 6 - 7 | 507 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RecQ like helicase 5 | RECQL5 | RECQ5 | -35.3kb | Anti-sense | Chr. 17 - 75626845 - 75667202 on NCBI Build GRCh38 |
Downstream | integrin subunit beta 4 | ITGB4 |
CD104, GP150 |
16.5kb | Sense | Chr. 17 - 75721328 - 75757819 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
