Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006673.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006673.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006673.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006673.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006673.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145509.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145509.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145509.hg.1 |
TC1300006673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145508.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145508.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145508.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145508.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145508.hg.1 |
TC1300006673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145507.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145507.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145507.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145507.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145507.hg.1 |
TC1300006673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006673.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006673.hg.1 |
ENST00000452222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006673.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006673.hg.1 |
NR_046548.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006673.hg.1 |
NR_046548.1 |
Refseq |