Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010197.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010197.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169607.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169607.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169607.hg.1 |
TC0600010197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600169606.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169606.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169606.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169606.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169606.hg.1 |
TC0600010197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600169604.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169604.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169604.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169604.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169604.hg.1 |
TC0600010197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010197.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010197.hg.1 |
ENST00000455390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010197.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169608.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169608.hg.1 |
NR_046644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169608.hg.1 |
TC0600010197.hg.1 |
Clariom D TC |