Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010511.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010511.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010511.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010511.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300176626.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300176626.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300176626.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300176626.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300176626.hg.1 |
TC0300010511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300176627.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300176627.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300176627.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300176627.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300176627.hg.1 |
TC0300010511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010511.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010511.hg.1 |
ENST00000426702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010511.hg.1 |
NR_046652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010511.hg.1 |
NR_046652.1 |
Refseq |