Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007813.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007813.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007813.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007813.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007813.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007813.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152665.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152665.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152665.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152665.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152665.hg.1 |
TC1300007813.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152666.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152666.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152666.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152666.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152666.hg.1 |
TC1300007813.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007813.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007813.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007813.hg.1 |
ENST00000439367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007813.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007813.hg.1 |
NR_046663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007813.hg.1 |
NR_046663.1 |
Refseq |