Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198332.hg.1 |
TC0300013171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198333.hg.1 |
TC0300013171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198334.hg.1 |
TC0300013171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000423873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
ENST00000457192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013171.hg.1 |
NR_046664.1 |
Refseq |