Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009892.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009892.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009892.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009892.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009892.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171904.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171904.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171904.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171904.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171904.hg.1 |
TC0300009892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300171905.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171905.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171905.hg.1 |
TC0300009892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009892.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009892.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009892.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009892.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009892.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171903.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171903.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171903.hg.1 |
ENST00000414634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171903.hg.1 |
NR_046758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171903.hg.1 |
TC0300009892.hg.1 |
Clariom D TC |