Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006485.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006485.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006485.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006485.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006485.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006485.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006485.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006485.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144632.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144632.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144632.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144632.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144632.hg.1 |
TC1200006485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200144633.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144633.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144633.hg.1 |
ENST00000541871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144633.hg.1 |
NR_046768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144633.hg.1 |
TC1200006485.hg.1 |
Clariom D TC |