Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205761.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205761.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205761.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205761.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205761.hg.1 |
TC0100014135.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014135.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205760.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205760.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205760.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205760.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205760.hg.1 |
TC0100014135.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205762.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205762.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205762.hg.1 |
ENST00000457061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205762.hg.1 |
NR_046839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205762.hg.1 |
TC0100014135.hg.1 |
Clariom D TC |