| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000455894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000455894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
ENST00000455894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000455894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000455894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
ENST00000455894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007346.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
ENST00000413402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149766.hg.1 |
TC1300007346.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300149769.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149769.hg.1 |
NR_047003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300149769.hg.1 |
TC1300007346.hg.1 |
Clariom D TC |