Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007820.hg.1 |
ENST00000562781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007820.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007820.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007820.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152688.hg.1 |
ENST00000562781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152688.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152688.hg.1 |
ENST00000562781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152688.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152688.hg.1 |
TC1300007820.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007820.hg.1 |
ENST00000562781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007820.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007820.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007820.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152686.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152686.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152686.hg.1 |
TC1300007820.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152687.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152687.hg.1 |
NR_047482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152687.hg.1 |
TC1300007820.hg.1 |
Clariom D TC |