Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007092.hg.1 |
ENST00000377927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007092.hg.1 |
ENST00000377927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007092.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007092.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148212.hg.1 |
ENST00000377927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148212.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148212.hg.1 |
ENST00000377927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148212.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148212.hg.1 |
TC0X00007092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007092.hg.1 |
ENST00000377927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007092.hg.1 |
ENST00000377927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007092.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007092.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148213.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148213.hg.1 |
NR_102268.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148213.hg.1 |
TC0X00007092.hg.1 |
Clariom D TC |