Assay Details
Gene Symbol: DNAH17-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102401.1 | 3 - 4 | 586 | 71 | ||
GenBank mRNA | BQ919011.1 | - | 1 - 2 | 64 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | phosphatidylglycerophosphate synthase 1 | PGS1 | -68.4kb | Sense | Chr. 17 - 78378618 - 78424659 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928710 | LOC101928710 | 123.2kb | Anti-sense | Chr. 17 - 78617384 - 78632057 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168478.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168479.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168489.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168489.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168489.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168489.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168489.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168486.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000588565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009054.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168487.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168483.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | ENST00000591373.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | ENST00000585969.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | ENST00000591373.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168475.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168484.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | ENST00000591373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168485.hg.1 | TC1700009054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700168488.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168488.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168488.hg.1 | ENST00000598378.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700168488.hg.1 | NR_102401.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700168488.hg.1 | TC1700009054.hg.1 | Clariom D TC |
