Assay Details
| Gene Symbol: LINC00678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102708.1 | 2 - 3 | 496 | 103 | ||
| GenBank mRNA | CD049808.1 | - | 2 - 3 | 464 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8087 | MIR8087 | hsa-mir-8087 | -103.1kb | Sense | Chr. 11 - 27514970 - 27515047 on NCBI Build GRCh38 |
| Downstream | brain derived neurotrophic factor | BDNF |
ANON2, BULN2 |
30.7kb | Sense | Chr. 11 - 27654893 - 27722058 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000527083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | ENST00000534757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010409.hg.1 | NR_102708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000419695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000419695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000419695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000454596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009462.hg.1 | ENST00000454596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006952.hg.1 | ENST00000555655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006952.hg.1 | ENST00000555655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006952.hg.1 | ENST00000555655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000519695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008910.hg.1 | ENST00000524275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000419695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000419695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000419695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000421310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000454596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009462.hg.1 | ENST00000454596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006952.hg.1 | ENST00000555655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006952.hg.1 | ENST00000555655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006952.hg.1 | ENST00000555655.1 | Ensembl |

LINC00678
Genomic Map