Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000514169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000514169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
ENST00000514169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192911.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192911.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192911.hg.1 |
TC0500013166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500192913.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192913.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192913.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192913.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192913.hg.1 |
TC0500013166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192918.hg.1 |
TC0500013166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000514169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000514169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
ENST00000514169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013166.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192912.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192912.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192912.hg.1 |
TC0500013166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
ENST00000503599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
ENST00000508795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192919.hg.1 |
TC0500013166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500192914.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192914.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192914.hg.1 |
ENST00000499046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192914.hg.1 |
NR_102752.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192914.hg.1 |
TC0500013166.hg.1 |
Clariom D TC |