Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179455.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179455.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179455.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179455.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179455.hg.1 |
TC1500010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500179458.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179458.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179458.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179458.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179458.hg.1 |
TC1500010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500179459.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179459.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179459.hg.1 |
TC1500010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000558722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
ENST00000560888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102753.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010563.hg.1 |
NR_102754.1 |
Refseq |