Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156252.hg.1 |
TC2100008293.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100156252.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156252.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156252.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156252.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156251.hg.1 |
TC2100008293.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100156251.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156251.hg.1 |
NR_103713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156251.hg.1 |
ENST00000450205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156251.hg.1 |
NR_103713.2 |
Refseq |