| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008293.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156249.hg.1 |
TC2100008293.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156249.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156249.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156250.hg.1 |
TC2100008293.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156250.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156250.hg.1 |
NR_103713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156250.hg.1 |
ENST00000450205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156250.hg.1 |
NR_103713.2 |
Refseq |