Assay Details
Gene Symbol: LINC00613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103763.1 | 2 - 3 | 129 | 159 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | poly(A) binding protein cytoplasmic 4 like | PABPC4L | -1.7mb | Sense | Chr. 4 - 133948459 - 134201748 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC729307 | LOC729307 | 1.3mb | Anti-sense | Chr. 4 - 137193756 - 137199948 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
