Assay Details
Gene Symbol: CCDC37-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103788.1 | 1 - 2 | 502 | 98 | ||
GenBank mRNA | AW241753.1 | - | 2 - 3 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Kruppel like factor 15 | KLF15 | KKLF | -36.1kb | Sense | Chr. 3 - 126288121 - 126357393 on NCBI Build GRCh38 |
Downstream | cilia and flagella associated protein 100 | CFAP100 |
CCDC37, MIA1 |
95.0 | Anti-sense | Chr. 3 - 126394893 - 126436556 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000505467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000505467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191855.hg.1 | TC0300012276.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000505467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000505467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012276.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | ENST00000505467.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | ENST00000505467.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191856.hg.1 | TC0300012276.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | ENST00000511287.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191854.hg.1 | TC0300012276.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191851.hg.1 | TC0300012276.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191857.hg.1 | TC0300012276.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | ENST00000506660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | NR_103787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | NR_103788.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | NR_103789.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300191858.hg.1 | TC0300012276.hg.1 | Clariom D TC |
