Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016440.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
ENST00000415396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221657.hg.1 |
TC0200016440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
ENST00000416466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221661.hg.1 |
TC0200016440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200221649.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221649.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221649.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221649.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221649.hg.1 |
TC0200016440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200221660.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221660.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221660.hg.1 |
ENST00000449601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221660.hg.1 |
NR_103805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221660.hg.1 |
TC0200016440.hg.1 |
Clariom D TC |