Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171436.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171436.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171436.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171436.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171436.hg.1 |
TC0800010513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171437.hg.1 |
TC0800010513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000517787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
ENST00000524136.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010513.hg.1 |
NR_103832.1 |
Refseq |