Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162608.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600162608.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162608.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600162608.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162608.hg.1 |
TC1600008308.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600162609.hg.1 |
TC1600008308.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
ENST00000561529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008308.hg.1 |
NR_103852.1 |
Refseq |