Assay Details
Gene Symbol: LINC01301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103854.1 | 2 - 3 | 401 | 118 | ||
GenBank mRNA | AK124262.1 | - | 2 - 3 | 402 | 118 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carbonic anhydrase 8 | CA8 |
CA-RP, CA-VIII, CALS, CAMRQ3, CARP |
-120.8kb | Sense | Chr. 8 - 60185420 - 60281423 on NCBI Build GRCh38 |
Downstream |
RAB2A, member RAS oncogene family |
RAB2A |
LHX, RAB2 |
108.4kb | Anti-sense | Chr. 8 - 60516910 - 60623644 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000532232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000532232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000532232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171869.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171869.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171869.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171869.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171869.hg.1 | TC0800010579.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800171870.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171870.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171870.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171870.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171870.hg.1 | TC0800010579.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171872.hg.1 | TC0800010579.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800171868.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171868.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171868.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171868.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171868.hg.1 | TC0800010579.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171871.hg.1 | TC0800010579.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000530725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000532232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000532232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000532232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | ENST00000534117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010579.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171867.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171867.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171867.hg.1 | TC0800010579.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | ENST00000476425.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | ENST00000506428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | ENST00000531654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | NR_103854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800171873.hg.1 | TC0800010579.hg.1 | Clariom D TC |
