| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000381475.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000417554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000417554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000417554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000440199.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000440199.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000440862.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000440862.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000440862.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000440862.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000442423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000442423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000442423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000613452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000613452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000613452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000613452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000614292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
ENST00000622328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_027468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018475.hg.1 |
NR_104014.2 |
Refseq |