Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011755.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011755.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011755.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011755.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011755.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011755.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011755.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011755.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180514.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180514.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180514.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180514.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180514.hg.1 |
TC1000011755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000180518.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180518.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180518.hg.1 |
ENST00000449805.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180518.hg.1 |
NR_104028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180518.hg.1 |
TC1000011755.hg.1 |
Clariom D TC |