Assay Details
| Gene Symbol: ATP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104109.1 | 2 - 3 | 324 | 159 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoglycerate mutase family member 4 | PGAM4 |
PGAM-B, PGAM1, PGAM3, dJ1000K24.1 |
-2.0kb | Anti-sense | Chr. X - 77967961 - 77969638 on NCBI Build GRCh38 |
| Downstream | phosphoglycerate kinase 1 | PGK1 |
HEL-S-68p, MIG10, PGKA |
72.6kb | Sense | Chr. X - 78104169 - 78126827 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ATP7A
Genomic Map